Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR MabSelectTM Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE28408253
Encompass
HiTrap(R) MabSelect(TM) columns are prepacked with MabSelect a Bioprocess medium for capuring of Mabs from large sample volumes.
Catalog No. 50-290-8140