Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR NHS-activated High
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17071601
Encompass
HiTrap(R) NHS activated HP columns are prepacked with NHS activated Sepharose(R) High Performance. This high-performance medium is designed for easy coupling of ligands containing primary amines.HiTrap(R) NHS-activated HP is a prepacked ready to use column for preparative affinity chromatography. The special design of the column together with the medium provide fast simple and easy separations in a convenient format. The column can be operated with a syringe peristaltic pump or liquid chromatography system such as AKTAdesign or fPLC(TM) System.
Catalog No. 50-290-4101