Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Protein A High Per
Catalog No. 502907397
Encompass
HiTrap(R) Protein A HP is a prepacked Protein A Sepharose(R) High Performance column for preparative purification of monoclonal and polyclonal antibodies from ascites serum and cell culture supernatants.HiTrap(R) Protein A HP is a prepacked ready to use column for preparative purification of monoclonal and polyclonal antibodies. The special design of the column together with the medium provide fast simple and easy separations in a convenient format.
Catalog No. 50-290-7397 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17040201
HiTrap(R) Protein A HP is a prepacked Protein A Sepharose(R) High Performance column for preparative purification of monoclonal and polyclonal antibodies from ascites serum and cell culture supernatants.HiTrap(R) Protein A HP is a prepacked ready to use column for preparative purification of monoclonal and polyclonal antibodies. The special design of the column together with the medium provide fast simple and easy separations in a convenient format.