Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Q fast flow Cytiva
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17505301
Encompass
HiTrap(R) Q ff is prepacked with Q Sepharose(R) fast flow and is a strong anion exchanger for small-scale protein purifications as well as screening of binding and elutions conditions.Q SP DEAE and CM Sepharose(R) fast flow are based on a robust 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose(R) 4 fast flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity which is particularly useful for the purification of high molecular mass proteins.
Catalog No. 50-290-1699