Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Q High Performance
Catalog No. 502904476
Encompass
HiTrap(R) Q HP are prepacked ready-to-use Q Sepharose(R) High Performance strong anion exchange columns for high-reSOLUTIONon small-scale protein purification.
Catalog No. 50-290-4476 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17115401
HiTrap(R) Q HP are prepacked ready-to-use Q Sepharose(R) High Performance strong anion exchange columns for high-reSOLUTIONon small-scale protein purification.