Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Q High Performance
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17115301
Encompass
HiTrap(R) Q HP are prepacked ready-to-use Q Sepharose(R) High Performance strong anion exchange columns for high-reSOLUTIONon small-scale protein purification.
Catalog No. 50-290-0393