Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR rprotein A Fast Fl
Catalog No. 502906954
Encompass
HiTrap(R) rProtein A FF columns are prepacked with rProtein A Sepharose(R) Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources including cell supernatants and ascites.The recombinant protein A ligand (produced in E. coli) is coupled to Sepharose(R) Fast Flow by a technique which generates a stable thioether linkage between rProtein A and the base matrix. The coupling technique is optimized to give high binding capacity for IgG.
Catalog No. 50-290-6954 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17508001
HiTrap(R) rProtein A FF columns are prepacked with rProtein A Sepharose(R) Fast Flow for purification and fractionation of IgG subclasses and IgG from different sources including cell supernatants and ascites.The recombinant protein A ligand (produced in E. coli) is coupled to Sepharose(R) Fast Flow by a technique which generates a stable thioether linkage between rProtein A and the base matrix. The coupling technique is optimized to give high binding capacity for IgG.