Learn More
Sigma Aldrich Fine Chemicals Biosciences HiTrapR Sp High Performanc
Catalog No. 502900379
Encompass
HiTrap(R) SP HP are prepacked ready-to-use SP Sepharose(R) High Performance strong cation exchange columns for high-reSOLUTIONon small-scale protein purification.The small particle size (34mum) of the medium allows fast adsorption and desorption even at high sample loadings and flow rates.SP Sepharose(R) High Performance is a strong cation exchange medium and Q Sepharose(R) High Performance is a strong anion exchange medium. Both remain charged and have high loading capacities over broad pH ranges.
Catalog No. 50-290-0379 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17115101
HiTrap(R) SP HP are prepacked ready-to-use SP Sepharose(R) High Performance strong cation exchange columns for high-reSOLUTIONon small-scale protein purification.The small particle size (34mum) of the medium allows fast adsorption and desorption even at high sample loadings and flow rates.SP Sepharose(R) High Performance is a strong cation exchange medium and Q Sepharose(R) High Performance is a strong anion exchange medium. Both remain charged and have high loading capacities over broad pH ranges.