Learn More
Bioss HIV-1
SDP
Supplier:  Bioss BSM72241M
Encompass_Preferred
HIV-1 Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications WB Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-64759