Learn More
CPC Scientific H-Ala-Val-Gly-Ile-Gly-Ala-Leu-Phe-Leu-Gly-Phe-Leu-Gly-Ala-Ala-Gly-Ser-Thr-Met-Gly-Ala-Arg-Ser-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV012B
Encompass
HIV envelope protein gp41 (519-541 (AVGIGALFLGFLGAAGSTMGARS-amide) destabilizes the lamellar structure of the membrane so that fusion can occur.ONE-LETTER SEQUENCE: AVGIGALFLGFLGAAGSTMGARS-NH2MOLECULAR FORMULA: C95H155N27O26S1MOLECULAR WEIGHT:2123.51STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [128631-86-7], RESEARCH AREA: AntimicrobialREFERENCES: J.L.Nieva et al., Lett. Pept. Sci., 4, 365 (1997); P.W.Mobley et al., Biochim. Biophys. Acta, 1418, 1 (1999); M.Kanyalkar et al., J. Pept. Sci., 10, 363 (2004)
Catalog No. 50-210-9552