Learn More
CPC Scientific H-Ser-Leu-Tyr-Asn-Thr-Val-Ala-Thr-Leu-OH (acetate salt) 5MG
SDP
Supplier:  CPC Scientific HIV014B
Encompass
HLA-A*0201-restricted immunodominant CD8 epitope of the HIV gag protein used for the characterization of CD8+ -T cells of HIV-positiv patients.ONE-LETTER SEQUENCE: SLYNTVATLMOLECULAR FORMULA: C44H72N10O15MOLECULAR WEIGHT:981.11STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [147468-65-3], RESEARCH AREA: AntimicrobialREFERENCES: D.Scott-Algara et al., J. Infect. Dis., 183, 1565 (2001); J.Wilkinson et al., J. Virol., 76, 2634 (2002); F.Buseyne et al., J. Virol., 76, 12414 (2002)
Catalog No. 50-210-9556