Learn More
CPC Scientific H-Ala-Met-Gln-Met-Leu-Lys-Asp-Thr-Ile-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV015A
Encompass
AMQMLKDTI represents a gag-specific MHC class I-restricted peptide.ONE-LETTER SEQUENCE: AMQMLKDTIMOLECULAR FORMULA: C44H79N11O14S2MOLECULAR WEIGHT:1050.31STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [334731-59-8], RESEARCH AREA: Antimicrobial
Catalog No. 50-210-9557