Learn More
ACROBiosystems Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade E (CM244)] GP120;HEK293;100UG;GP4
Catalog No. 501017912
Encompass_Preferred
Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade E (CM244)] GP120;100UG;HIV-1 [HIV-1 / Clade E (CM244)] GP120 (GP4-V15223) is expressed from human 293 cells (HEK293). It contains AA Thr 36 - Lys 511 (Accession # Q4QX31).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.1 kDa. The protein migrates as 65-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 20-156 ng/mL (QC tested).;Immobilized Biotinylated Human CD4, Fc Tag (Cat. No. CD4-H82F3) at 1 μg/mL (100 μL/well) can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 0.156-0.313 μg/mL (Routinely tested).
Catalog No. 50-101-7912 Supplier ACROBiosystems Supplier No. GP4V15223100UG
Recombinant Protein;Recombinant HIV-1 [HIV-1/Clade E (CM244)] GP120;100UG;HIV-1 [HIV-1 / Clade E (CM244)] GP120 (GP4-V15223) is expressed from human 293 cells (HEK293). It contains AA Thr 36 - Lys 511 (Accession # Q4QX31).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 54.1 kDa. The protein migrates as 65-110 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD4, Fc Tag (Cat. No. CD4-H5259) at 5 μg/mL (100 μL/well) can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 20-156 ng/mL (QC tested).;Immobilized Biotinylated Human CD4, Fc Tag (Cat. No. CD4-H82F3) at 1 μg/mL (100 μL/well) can bind HIV-1 [HIV-1 / Clade E (CM244)] GP120 (Cat. No. GP4-V15223) with a linear range of 0.156-0.313 μg/mL (Routinely tested).