Learn More
Creative Biomart Recombinant HIV-1 gp41 Antigen, b-galactosidase-tagged, Biotin-Labled
Supplier:  Creative Biomart HIV269
Encompass_Preferred
HIV-1 gp41 Biotin labeled is a non-glycosylated polypeptide chain, containing 288 amino acids (466-753 a.a.) and having an Mw of 32kDa. HIV1 gp41 biotin labeled is fused to an 114 kDa beta-galactosida
Catalog No. 50-208-4647