Learn More
Bioworld HIV-1 GP41 LONG (513-674 A.
Supplier:  Bioworld 220602051
Encompass_Preferred
HIV-1 GP41 LONG (513-674 A.A.), (RECOMBINANT): Recombinant HIV-1 gp41 Long Source:E.coli Description : The protein contains the full-length sequence of HIV-I glycoprotein gp41L. Purification method: Purified by proprietary chromato...
Catalog No. 50-253-306