Learn More
Creative Biomart Recombinant HIV-1 gp41 Long Antigen, Biotin-Labled
Supplier:  Creative Biomart HIV267
Encompass_Preferred
The E. coli derived 32 kDa biotin labeled recombinant protein is a non-glycosylated polypeptide chain, containing the HIV-1 p41 immunodominant regions, amino acids 466-753. The HIV-1 p41 Biotinylated
Catalog No. 50-208-4645