Learn More
Bioworld HIV-1 GP41 (RECOMBINANT)
Supplier:  Bioworld 220602021
Encompass_Preferred
Recombinant HIV-1 gp41 Description : HIV-1 env gp41 Strain IIIB is a non-glycosylated 288 amino acids polypeptide chain (a.a. 466-753) having a molecular mass of 32kDa. The protein is fused to B-galactosidase (114 kDa) at the N- ter...
Catalog No. 50-253-303