Learn More
Medchemexpress LLC HIV-1 inhibitor-45 | 2677762-43-3 | C23H24N4O8S | 5 MG
SDP
Catalog No. 5000375464
Encompass_Preferred
HIV-1 inhibitor-45 is a potent HIV-1 RNase H inhibitor with an IC50 value of 0.067 μM. It exhibits antiviral activity but shows poor cell permeability.
  • Potent HIV-1 RNase H inhibitor
  • IC50 value of 0.067 μM
  • Shows antiviral activity
  • Poor cell permeability (apparent permeability coefficient (Papp) lower than 0.48 x 10-6 cm/s)
  • Low toxicity with a CC50 value of 24.7 μM in MT-4 cells
  • Poor membrane permeability attributed to its polyphenolic skeleton
Catalog No. 50-003-75464 Supplier Medchemexpress LLC Supplier No. HY1507595MG
HIV-1 inhibitor-45 is a potent HIV-1 RNase H inhibitor with an IC50 value of 0.067 μM. It exhibits antiviral activity but shows poor cell permeability.
  • Potent HIV-1 RNase H inhibitor
  • IC50 value of 0.067 μM
  • Shows antiviral activity
  • Poor cell permeability (apparent permeability coefficient (Papp) lower than 0.48 x 10-6 cm/s)
  • Low toxicity with a CC50 value of 24.7 μM in MT-4 cells
  • Poor membrane permeability attributed to its polyphenolic skeleton