Learn More
Creative Biomart Influenza A virus H3N2 (Kiev) Antigen
Catalog No. 502084650
Encompass_Preferred
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Kiev/301/94 like /Johannesburg/33/94. The Influenza Virus was purified by Ultracentrifugation with 10-40 %
Catalog No. 50-208-4650 Supplier Creative Biomart Supplier No. HIV272
Allantoic fluid of 10 days old embryonated eggs, inoculated with influenza A virus, strain A/Kiev/301/94 like /Johannesburg/33/94. The Influenza Virus was purified by Ultracentrifugation with 10-40 %