Learn More
Creative Biomart Recombinant HIV-1 p24 Antigen, GST-tagged, HRP-Labled
Supplier:  Creative Biomart HIV282
Encompass_Preferred
The E. coli derived HRP recombinant protein is a non-glycosylated polypeptide chain, containing the HIV-1 p24 immunodominant regions and fused to a GST tag at N-terminus.
Catalog No. 50-208-4660