Learn More
Bioworld HIV-1 pol Integrase (Recombinant), 100 μg
Supplier:  Bioworld 220602161
Encompass_Preferred
Recombinant HIV-I pol Integrase Source: E. coli Description : The E.coli derived 26 kDa recombinant protein is a non-glycosylated polypeptide chain, containing the HIV-1 immunodominant regions from the pol protein (intergrase) and fused with six histidines at the N-terminus.Purity: Protein is >95% pure as determined by HPLC analysis and SDS PAGE. Formulation: 4.55mg/ml 1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X & 50% Glycerol. Stability: HIV-1 Integrase although stable at 4°C for 1 week, should be stored below -18°C. Please prevent freeze thaw cycle.Specificity: Immunoreactive with all sera of HIV-I infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for early detection of HIV seroconvertors with minimal specificity problems. Usage: BioWORLD's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-317
 Restricted Item