Learn More
CPC Scientific H-Thr-Arg-Gln-Ala-Arg-Arg-Asn-Arg-Arg-Arg-Arg-Trp-Arg-Glu-Arg-Gln-Arg-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIV018A
Encompass
This arginine-rich fragment interacts specifically with RNA. It has been shown that rev protein and rev protein (34-50) bind IIB RNA with a similar dissociation constant of approx. 10 nM.ONE-LETTER SEQUENCE: TRQARRNRRRRWRERQRMOLECULAR FORMULA: C97H173N51O24MOLECULAR WEIGHT:2437.77STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [141237-50-5], RESEARCH AREA: AntimicrobialREFERENCES: B.J.Calnan et al., Genes Dev., 5, 201 (1991); J.Kjems et al., EMBO J., 11, 1119 (1992)
Catalog No. 50-210-9563