Learn More
Medchemexpress LLC HY-P1491 1mg Medchemexpress, HIV-1 TAT (48-60) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP14911MG
Encompass_Preferred
Medchemexpress, HY-P1491 1mg HIV-1 TAT (48-60) CAS: HIV-1 TAT (48-60) is a cell-penetrating peptide derived from the human immunodeficient virus (HIV)-1 Tat protein residue 48-60. It has been used to deliver exogenous macromolecules into cells in a non-disruptive way. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3061