Learn More
Creative Biomart Recombinant Parvovirus B19 VLP VP1/VP2 Antigen
Supplier:  Creative Biomart HIV286
Encompass_Preferred
Recombinant Parvovirus B19 VLP VP1/VP2 Co-Capsid produced in SF9 containing B19 capsid proteins VP1 (81 kDa) and VP2 (61 kDa).Parvovirus B19 VLP VP1/VP2 is purified by proprietary chromatographic tech
Catalog No. 50-208-4664