Learn More
Bioworld HIV-1 TAT CLADE-B (RECOMBIN
Supplier:  Bioworld 220602171
Encompass_Preferred
HIV-1 TAT CLADE-B (RECOMBINANT): Description :Recombinant HIV-I TAT produced in E.coli is a single, non-glycosylated, polypeptide chain containing 86 aminoacids encoded by two exons and having chain having a molecular mass of 14kDa. Sour...
Catalog No. 50-253-318