Learn More
Bachem HIV-1 tat Protein (47-57) Trifluoroacetate
SDP
Supplier:  Bachem 4048761.0005
Encompass_Preferred
HIV-1 tat Protein (47-57) Trifluoroacetate, H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-OH, 191936-91-1, C64H118N32O14, Mr 1559.85, 5mg. YGRKKRRQRRR, a cell-permeable peptide (CPP) derived from the HIV-1 tat protein transduction domain (PTD), has been shown to promote the entry of nucleic acids into several cell types.
Catalog No. 50-260-054