Learn More
CPC Scientific H-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HIV020A
Encompass
RKKRRQRRR, a nine amino acid motif within the HIV-1 tat protein transduction domain (PTD), suffices for the transduction of heterologous proteins through the plasma membrane.ONE-LETTER SEQUENCE: RKKRRQRRRMOLECULAR FORMULA: C53H106N30O11MOLECULAR WEIGHT:1339.62STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123251-89-8], RESEARCH AREA: AntimicrobialREFERENCES: J.Park et al., J. Gen. Virol., 83, 1173 (2002)
Catalog No. 50-210-9567