Learn More
Bioworld HIV-2 gp36 (390-702) (Recombinant), 100 μg
Supplier:  Bioworld 220602221
Encompass_Preferred
Recombinant HIV-II coat protein gp36 fragment Source: E.coli Description : The protein contains the a fragment of HIV-II coat protein gp36 ,amino acids:(390-702). MW=34 kD. Purification method: Purified by proprietary chromatographic technique. Purity: Protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. Protein conc: 1mg/ml, 20mM PBS pH 7.8, NaCl 0.5M, DTT 1mM, 8M urea and 0.4M imidasole. Storage: Protein is shipped at ambient temperature. Upon arrival, Store at -20C. Stability: Five years frozen. One month in solution at room temperature. Specificity: Immunoreactive with all sera of HIV-I infected individuals. Application: Antigen in ELISA and Western blots, excellent antigen for early detection of HIV seroconvertors with minimal specificity problems. Usage: BioWORLD\'s products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or household chemicals.
Catalog No. 50-253-323
 Restricted Item