Learn More
CPC Scientific H-Asn-Asn-Thr-Arg-Lys-Ser-Ile-Arg-Ile-Gln-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-Gly-Lys-Ile-Gly-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV007B
Encompass
This peptide derived from the V3 loop of HIV IIIB gp120 completely blocks the fusion inhibition activity of antisera to recombinant human HIV proteins. NNTRKSIRIQRGPGRAFVTIGKIG also inhibits the infection of primary human T cells by HIV-1 IIIB, as well as syncytium formation.ONE-LETTER SEQUENCE: NNTRKSIRIQRGPGRAFVTIGKIGMOLECULAR FORMULA: C114H199N41O31MOLECULAR WEIGHT:2640.06STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [115416-08-5], SYNONYMS: gp120 Fragment 308-331, HIV (gp120) Fragment (308-331);RESEARCH AREA: AntimicrobialREFERENCES: L.Ratner et al., Nature, 313, 277 (1985); J.R.Rusche et al., Proc. Natl. Acad. Sci. USA, 85, 3198 (1988); P.N.Nehete et al., J. Virol., 67, 6841 (1993)
Catalog No. 50-210-9542