Learn More
CPC Scientific H-Cys-Thr-His-Gly-Ile-Arg-Pro-Val-Val-Ser-Thr-Gln-Leu-Leu-Leu-Asn-Gly-Ser-Leu-Ala-Glu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV006B
Encompass
This fragment with sequence homology to a domain of the external envelope glycoprotein (gp120) of the human immunodeficiency virus (HIV) is important for HIV infectivity and antibody neutralization.ONE-LETTER SEQUENCE: CTHGIRPVVSTQLLLNGSLAEMOLECULAR FORMULA: C95H162N28O30S1MOLECULAR WEIGHT:2208.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [113944-46-0], RESEARCH AREA: AntimicrobialREFERENCES: D.D.Ho et al., Science, 239, 1021 (1988)
Catalog No. 50-210-9540