Learn More
CPC Scientific H-Ala-Val-Gly-Ile-Gly-Ala-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HIV011A
Encompass
The hexapeptide AVGIGA is identical in sequence to the first 6 N-terminal amino acids of the gp41 subunit of the HIV envelope glycoprotein. It was found to completely inhibit the HIV-induced syncytium formation (ID50 = 0.75 mM).SEQUENCE: H-Ala-Val-Gly-Ile-Gly-Ala-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: AVGIGAMOLECULAR FORMULA: C21H38N6O7MOLECULAR WEIGHT: 486.57STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [129426-47-7]SYNONYMS: RESEARCH AREA: AntimicrobialREFERENCES:SKU(s): HIV-011A, HIV-011B
Catalog No. 50-210-9549