Learn More
CPC Scientific H-Arg-Gly-Pro-Gly-Arg-Ala-Phe-Val-Thr-Ile-OH (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific HIV008B
Encompass
The decapeptide RGPGRAFVTI, derived from the V3 loop region of HIV-1 type IIIB gp120, efficiently induced HIV-1 env-specific cytotoxic T lymphocyte activity in mice (no carrier needed).ONE-LETTER SEQUENCE: RGPGRAFVTIMOLECULAR FORMULA: C48H80N16O12MOLECULAR WEIGHT:1073.25STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [147841-68-7], RESEARCH AREA: AntimicrobialREFERENCES: P.N.Nehete et al., Cell. Immunol., 160, 217 (1995); Y.Nakagawa et al., Immunology, 101, 76 (2000)
Catalog No. 50-210-9544