Learn More
CPC Scientific H-Lys-Gln-Phe-Ile-Asn-Met-Trp-Gln-Glu-Val-Gly-Lys-Ala-Met-Tyr-Ala-Pro-Pro-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV009B
Encompass
The octadecapeptide derived from the CD4 attachment region of HIV gp120 inhibited the syncytial formation in vitro. According to immunological studies, the HIV (gp120) fragment 421-438 generated high titres of anti-gp120 antibodies following immunization in mice, rabbits, and goats.ONE-LETTER SEQUENCE: KQFINMWQEVGKAMYAPPMOLECULAR FORMULA: C99H148N24O25S2MOLECULAR WEIGHT:2138.54STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [129318-38-3], RESEARCH AREA: AntimicrobialREFERENCES: W.J.W.Morrow et al., Immunology, 75, 557 (1992); R.M.Ferraz et al., J. Mol. Recognit., 22, 255 (2009)
Catalog No. 50-210-9546