Learn More
Sino Biological HIV Gag-p24 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502373322
Encompass_Preferred
Each vial contains 24 ng of recombinant Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 1200 pg/mL is recommended.
Catalog No. 50-237-3322 Supplier Sino Biological Supplier No. SEK1169515
Each vial contains 24 ng of recombinant Human Immunodeficiency Virus type 1 (HIV-1) p24 / Capsid Protein p24. Reconstitute with 1 mL Dilution Buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in Dilution Buffer, and a high standard of 1200 pg/mL is recommended.