Learn More
Sino Biological HIV Gag-p24 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11695R00220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant HIV-1 Gag-P24 (rv Gag-P24; Catalog#11695-V08E; ACI05538.1; Pro 133-Leu 363).
Catalog No. 50-237-1658