Learn More
Sino Biological HIV Gag-p24 PAb 20ul (Sino Biological)
Supplier:  Sino Biological 11695RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 Gag-P24 (Catalog#11695-V08E; ACI05538.1; Pro 133-Leu 363). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3009