Learn More
Sino Biological HIV-1 p24 Protein (group M, subtype D, strain NDK) Antibody, Rabbit PAb
Supplier:  Sino Biological 40252RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 p24 Protein (group M, subtype D, strain NDK) (Catalog#40252-V08E; AAA44868.1; Pro133-Leu363). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8416