Learn More
Sino Biological HIV-1 p24 (group N, strain 06CM-U14296) Antibody, Rabbit PAb
Supplier:  Sino Biological 40254RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 p24 (group N, strain 06CM-U14296) (Catalog#40254-V08E; Pro133-Leu363). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1406