Learn More
Sino Biological HIV-1 p24 (group M, subtype B, strain HXB2) Antibody, Rabbit Pab, Antigen Affinity Purified
Supplier:  Sino Biological 40243T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 p24 (group M, subtype B, strain HXB2) (Catalog#40243-V08E; AAB50258.1; Pro133-Leu363). HIV-1 p24 (group M, subtype B, strain HXB2) specific IgG was purified by HIV-1 p24 (group M, subtype B, strain HXB2) affinity chromatography.
Catalog No. 50-240-8792