Learn More
Sino Biological HIV-1 p24 (group M, subtype D, strain NDK) Antibody, Rabbit Pab, Antigen Affinity Purified
Supplier:  Sino Biological 40252T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 p24 (group M, subtype D, strain NDK) ( Catalog#40252-V08E; AAA44868.1; Pro133-Leu363). HIV-1 p24 (group M, subtype D, strain NDK) specific IgG was purified by HIV-1 p24 (group M, subtype D, strain NDK) affinity chromatography.
Catalog No. 50-240-9912