Learn More
Sino Biological HIV-1 p24 protein (group N, strain 06CM-U14296) Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40254T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 p24 protein (group N, strain 06CM-U14296) (Catalog#40254-V08E; Pro133-Leu363). HIV-1 p24 protein (group N, strain 06CM-U14296) specific IgG was purified by HIV-1 p24 protein (group N, strain 06CM-U14296) affinity chromatography.
Catalog No. 50-240-8538