Learn More
Sino Biological HIV-1 (group M, subtype B, strain 92418) p24 Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11695T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 (group M, subtype B, strain 92418) p24 ( Catalog#11695-V08E; ACI05538.1; Pro133-Leu363). HIV-1 (group M, subtype B, strain 92418) p24 specific IgG was purified by HIV-1 (group M, subtype B, strain 92418) p24 affinity chromatography.
Catalog No. 50-241-1345