Learn More
Sino Biological HIV-1 p24 Protein (group M, subtype B, strain HXB2) Antibody, Rabbit PAb
Supplier:  Sino Biological 40243RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 p24 Protein (group M, subtype B, strain HXB2) (Catalog#40243-V08E; AAB50258.1; Pro133-Leu363). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0888