Learn More
Sino Biological Human Immunodeficiency Virus type 1 (HIV-1) Gag-p24 Antibody (HRP), Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 11695RP02H20
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 Gag-P24 (Catalog#11695-V08E; ACI05538.1; Pro 133-Leu 363). Gag-P24 specific IgG was purified by HIV-1 Gag-P24 affinity chromatography, and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-4604