Learn More
Sino Biological HIV gp120 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502370745
Encompass_Preferred
Each vial contains 168 ng of recombinant Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 3000 pg/mL is recommended.
Catalog No. 50-237-0745 Supplier Sino Biological Supplier No. SEK1123315
Each vial contains 168 ng of recombinant Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120. Reconstitute with 1 mL dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in dilution buffer, and a high standard of 3000 pg/mL is recommended.