Learn More
Sino Biological HIV gp120 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11233R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant HIV-1 gp120 (group M, subtype CRF07_BC) ( Catalog#11233-V08H; AFM77980.1;Ala29-Glu502).
Catalog No. 50-237-2076