Learn More
Sino Biological HIV gp120 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11233RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HIV-1 gp120 (rv GP120; Catalog#11233-V08H;Ala29-Glu502; AFM77980.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-3002