Learn More
Sino Biological Human immunodeficiency virus HIV gp120 Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 40405V49HB20
Encompass_Preferred
A DNA sequence encoding the HIV gp120 (AAC31819.1) (Thr30-Arg513) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-3720