Learn More
XpressBio HIV-1 Integrase Wild-Type Assay
SDP
Catalog No. NC1345891
Encompass_Preferred
The HIV-1 Integrase Assay Kit is a non-radioactive assay used to quantitatively measure integrase activity, the effects of interacting proteins, anti-viral compounds, and other test articles on HIV-1 integrase activity.
Catalog No. NC1345891 Supplier XpressBio Supplier No. EZ1700
The HIV-1 Integrase Assay Kit is a non-radioactive assay used to quantitatively measure integrase activity, the effects of interacting proteins, anti-viral compounds, and other test articles on HIV-1 integrase activity.