Learn More
Bioss HIV TAT specific factor 1 Reco
SDP
Catalog No. 5000067355
Encompass_Preferred
HIV TAT specific factor 1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IHC-P IHC-F IF(ICC) IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-67355 Supplier Bioss Supplier No. BSM62353R
HIV TAT specific factor 1 Primary Antibody Unconjugated Host Species Rabbit Target Species Human Mouse Clonality Monoclonal Applications WB IHC-P IHC-F IF(ICC) IP IF RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol